MilliporeSigma
  • A New Method for In-Situ Skin Penetration Analysis by Confocal Raman Microscopy.
A New Method for In-Situ Skin Penetration Analysis by Confocal Raman Microscopy.
Molecules (Basel, Switzerland) (2020-09-19)
Richard Krombholz, Dominique Lunter
ABSTRACT
In the development of dermal drug formulations and cosmetics, understanding the penetration properties of the active ingredients is crucial. Given that widespread methods, including tape stripping, lack in spatial resolution, while being time- and labour-intensive, Confocal Raman Microscopy is a promising alternative. In optimizing topically applied formulations, or the development of generic formulations, comparative in-situ measurements have a huge potential of saving time and resources. In this work, we show our approach to in-situ skin penetration analysis by confocal Raman Microscopy. To analyse feasibility of the approach, we used caffeine solutions as model vehicles and tested the effectiveness of 1,2-pentanediol as a penetration enhancer for delivery to the skin.
Sigma-Aldrich
1,2-Pentanediol, 96%