MilliporeSigma
  • Use of new scratch test and tensile test for evaluation of bond strength of selfadhesive flowable resin composite for repair of artificial tooth erosion.
Use of new scratch test and tensile test for evaluation of bond strength of selfadhesive flowable resin composite for repair of artificial tooth erosion.
Dental materials journal (2020-01-15)
Yuki Murase, Hirotomo Kotake, Shusuke Kusakabe, Katsushi Okuyama, Yukimichi Tamaki, Masato Hotta
ABSTRACT
The purpose of this study was to use a new scratch test and tensile test to evaluate the bond strength between artificial erosive enamel or dentin and self-adhesive resin composites as a coating material. Coronal enamel or dentin surface was exposed to an erosive cycle (artificial saliva [AS], pH:7.0 for 6.5 h and acidic carbonated beverages for 5 min, alternated 3 times per day) for the eroded-surface or stored in AS for the remineralized-surface. Two self-adhesive flowable resin composites, Fusio and LLB-CR6 (prototype), and a conventional flowable resin composite, BEAUTIFIL FLOW with self-etching primer system, Clearfil Mega Bond, were applied to enamel or dentin surfaces; and then the bond strengths were measured. For the eroded-surface, there were no significant differences in bonding strength among all materials, as assessed by the new scratch test. Thus, these self-adhesive flowable resin composites might be useful for coating materials on acid-eroded tooth surfaces.
Sigma-Aldrich
1,2,4-Benzenetricarboxylic acid, ≥99%