MilliporeSigma
  • Mechanically strengthened hybrid peptide-polyester hydrogel and potential applications in spinal cord injury repair.
Mechanically strengthened hybrid peptide-polyester hydrogel and potential applications in spinal cord injury repair.
Biomedical materials (Bristol, England) (2020-06-20)
Hong Zhai, Jing Zhou, Jinghui Xu, Xiumin Sun, Yiwei Xu, Xibo Qiu, Chi Zhang, Zejia Wu, Houqing Long, Ying Bai, Daping Quan
ABSTRACT
ADA16 peptide hydrogels have been broadly used in tissue engineering due to their good biocompatibility and nanofibrous structure mimicking the native extracellular matrix (ECM). However, the low mechanical strength often fails them as implantable scaffolds. To improve the mechanical stability of the RADA16 peptide hydrogel, a photocrosslinkable diacrylated poly(ϵ-caprolactone)-b-poly(ethylene glycol)-b-poly(ϵ-caprolactone) triblock copolymer (PCECDA) was physically combined with RADA16 peptide pre-modified with cell adhesive Arg-Gly-Asp sequence (RADA16-RGD). Consequently, an interpenetrating network, RADA16-RGD/PCECDA, was formed with highly enhanced mechanical property. The storage modulus (G') of RADA16-RGD/PCECDA (6% w/v, mass ratio mRADA16-RGD/mPCECDA = 1:5) hybrid hydrogel was elevated to ∼2000 Pa, compared to the RADA16-RGD (1% w/v) hydrogel alone (∼700 Pa). Furthermore, this hybrid hydrogel retained the nanofibrous structure from RADA16-RGD peptide, but underwent much slower degradation than RADA16-RGD alone. In vitro, the hybrid hydrogel exhibited excellent cytocompatibility and promoted the differentiation of the seeded neural stem cells. Finally, the RADA16-RGD/PCECDA hydrogel demonstrated capability in reducing cavitation, glial scar formation and inflammation at the lesion sites of hemi-sectioned spinal cord injury model in rats, which holds great potential for application in neural tissue engineering and regenerative medicine.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Neurotrophin-3 human, recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Human NT-3 ELISA Kit, for serum, plasma, cell culture supernatant and urine