MilliporeSigma
  • Oroxylin A suppresses ACTN1 expression to inactivate cancer-associated fibroblasts and restrain breast cancer metastasis.
Oroxylin A suppresses ACTN1 expression to inactivate cancer-associated fibroblasts and restrain breast cancer metastasis.
Pharmacological research (2020-06-04)
Yue Cao, Wangjia Cao, Yangmin Qiu, Yuxin Zhou, Qinglong Guo, Yuan Gao, Na Lu
ABSTRACT
Tumor initiation and progression are not only ascribed to the behavior of cancer cells, but also profoundly influenced by the tumor microenvironment. Inside, cancer-associated fibroblasts (CAFs) have become key factors to accelerate growth and metastasis for the abundance in most solid tumors. Our group previously reported that Oroxylin A (OA), a flavone from Scutellaria Baicalensis Georgi, possess the ability to suppress growth and invasion of several tumor cells. However, the regulatory effect of OA on stromal microenvironment is poorly understood. In this study, breast cancer-induced fibroblasts and primary breast CAFs from MMTV-PyMT mice were used to evaluate the influence of OA on the activation of fibroblasts. Results showed that OA could decrease the expression of α-SMA, fibronectin, vimentin and matrix metalloproteinases (MMPs). Thus, OA-deactivated CAFs did not further promote the proliferation and invasion in breast cancer cells. In vivo experiments, OA could also impede tumor metastasis through exhausting progressive CAFs. Mechanically, OA could specifically bind ACTN1 and significantly inhibit its expression to prevent CAF activation. As a consequence, OA could decrease the phosphorylation of FAK and STAT3, and reduce the secretion of CCL2 in CAFs. Altogether, OA could remodel stromal microenvironment and it is a potential therapeutic agent in breast cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human ACTN1