MilliporeSigma
  • Inhibition of Porphyromonas gulae and periodontal disease in dogs by a combination of clindamycin and interferon alpha.
Inhibition of Porphyromonas gulae and periodontal disease in dogs by a combination of clindamycin and interferon alpha.
Scientific reports (2020-02-23)
Ryota Nomura, Hiroaki Inaba, Hidemi Yasuda, Mitsuyuki Shirai, Yukio Kato, Masaru Murakami, Naoki Iwashita, So Shirahata, Sho Yoshida, Saaya Matayoshi, Junya Yasuda, Nobuaki Arai, Fumitoshi Asai, Michiyo Matsumoto-Nakano, Kazuhiko Nakano
ABSTRACT
Porphyromonas gulae is a major periodontal pathogen in dogs, which can be transmitted to their owners. A major virulence factor of P. gulae consists of a 41-kDa filamentous appendage (FimA) on the cell surface, which is classified into three genotypes: A, B, and C. Thus far, inhibition of periodontal disease in dogs remains difficult. The present study assessed the inhibitory effects of a combination of clindamycin and interferon alpha (IFN-α) formulation against P. gulae and periodontal disease. Growth of P. gulae was significantly inhibited by clindamycin; this inhibition had a greater effect on type C P. gulae than on type A and B isolates. In contrast, the IFN-α formulation inhibited the expression of IL-1β and COX-2 elicited by type A and B isolates, but not that elicited by type C isolates. Furthermore, periodontal recovery was promoted by the administration of both clindamycin and IFN-α formulation to dogs undergoing periodontal treatment; moreover, this combined treatment reduced the number of FimA genotypes in oral specimens from treated dogs. These results suggest that a combination of clindamycin and IFN-α formulation inhibit P. gulae virulence and thus may be effective for the prevention of periodontal disease induced by P. gulae.
Sigma-Aldrich
Human COX-2 ELISA Kit, for cell culture supernatants, plasma, and serum samples
Pricing and availability is not currently available.