MilliporeSigma
  • Baicalin-Copper Complex Modulates Gut Microbiota, Inflammatory Responses, and Hormone Secretion in DON-Challenged Piglets.
Baicalin-Copper Complex Modulates Gut Microbiota, Inflammatory Responses, and Hormone Secretion in DON-Challenged Piglets.
Animals : an open access journal from MDPI (2020-09-04)
Andong Zha, Zhijuan Cui, Ming Qi, Simeng Liao, Jia Yin, Bie Tan, Peng Liao
ABSTRACT
The present experiment assessed the inflammatory responses, hormone secretion, and gut microbiota of weanling piglets administered baicalin-copper complex (BCU) or deoxynivalenol (DON) supplementation diets. Twenty-eight piglets were randomly assigned to four groups: control diet (Con group), a 4 mg DON/kg diet (DON group), a 5 g BCU/kg diet (BCU group), a 5 g BCU + 4 mg DON/kg diet (DBCU group). After 14 days, the results showed that dietary BCU supplementation remarkably increased the relative abundance of Clostrium bornimense and decreased the relative abundance of Lactobacillus in the DBCU group (p < 0.05). BCU decreased the serum concentration of IgG, IL-2, IFN-γ, and IgA in DON treated piglets (p < 0.05), and promoted the serum concentration of IL-1β, IgG, IL-2, IFN-γ, IgA, IL-6, IgM, and TNFα in normal piglets (p < 0.05). BCU increased the concentrations of serum IGF1, insulin, NPY, GLP-1, and GH, and decreased the concentrations of serum somatostatin in no DON treated piglets (p < 0.05). Dietary BCU supplementation significantly promoted the secretion of somatostatin, and inhibited the secretion of leptin in piglets challenged with DON (p < 0.05). BCU regulated the expression of food intake-related genes in the hypothalamus and pituitary of piglets. Collectively, dietary BCU supplementation alleviated inflammatory responses and regulated the secretion of appetite-regulating hormones and growth-axis hormones in DON challenged piglets, which was closely linked to changes of intestinal microbes.
Sigma-Aldrich
Human AGRP / Agouti-related Protein ELISA Kit, for serum, plasma, cell culture supernatants and urine