MilliporeSigma
  • Intracellular pH and free calcium changes in single cells using quene 1 and quin 2 probes and fluorescence microscopy.
Intracellular pH and free calcium changes in single cells using quene 1 and quin 2 probes and fluorescence microscopy.
FEBS letters (1983-09-05)
J Rogers, T R Hesketh, G A Smith, M A Beaven, J C Metcalfe, P Johnson, P B Garland
ABSTRACT
Photometric fluorescence microscopy has been used to measure intracellular pH (pHi) and free calcium concentrations [( Ca]i) in individual mouse thymocytes and 2H3 rat basophil leukaemic cells containing indicators for pH (quene 1) or calcium (quin 2). The pHi and [Ca]i measurements in individual 2H3 cells and mouse thymocytes and their responses to various stimuli were consistent with the corresponding data obtained from suspensions of these cells measured in a spectrofluorimeter. Photometric fluorescence microscopy of these indicators in individual cells provides a sensitive and fast method of following pHi and [Ca]i responses in individual cells.