MilliporeSigma
Platelet-derived serotonin links vascular disease and tissue fibrosis.
The Journal of experimental medicine (2011-04-27)
Clara Dees, Alfiya Akhmetshina, Pawel Zerr, Nicole Reich, Katrin Palumbo, Angelika Horn, Astrid Jüngel, Christian Beyer, Gerhard Krönke, Jochen Zwerina, Rudolf Reiter, Natalia Alenina, Luc Maroteaux, Steffen Gay, Georg Schett, Oliver Distler, Jörg H W Distler
ABSTRACT
Vascular damage and platelet activation are associated with tissue remodeling in diseases such as systemic sclerosis, but the molecular mechanisms underlying this association have not been identified. In this study, we show that serotonin (5-hydroxytryptamine [5-HT]) stored in platelets strongly induces extracellular matrix synthesis in interstitial fibroblasts via activation of 5-HT(2B) receptors (5-HT(2B)) in a transforming growth factor β (TGF-β)-dependent manner. Dermal fibrosis was reduced in 5-HT(2B)(-/-) mice using both inducible and genetic models of fibrosis. Pharmacologic inactivation of 5-HT(2B) also effectively prevented the onset of experimental fibrosis and ameliorated established fibrosis. Moreover, inhibition of platelet activation prevented fibrosis in different models of skin fibrosis. Consistently, mice deficient for TPH1, the rate-limiting enzyme for 5-HT production outside the central nervous system, showed reduced experimental skin fibrosis. These findings suggest that 5-HT/5-HT(2B) signaling links vascular damage and platelet activation to tissue remodeling and identify 5-HT(2B) as a novel therapeutic target to treat fibrotic diseases.
Sigma-Aldrich
Trichrome Stain (Masson) Kit
Pricing and availability is not currently available.