MilliporeSigma
  • Effect of the Trifunctional Chain Extender on Intrinsic Viscosity, Crystallization Behavior, and Mechanical Properties of Poly(Ethylene Terephthalate).
Effect of the Trifunctional Chain Extender on Intrinsic Viscosity, Crystallization Behavior, and Mechanical Properties of Poly(Ethylene Terephthalate).
ACS omega (2020-08-11)
Zhiyuan Zhao, Yinqiu Wu, Kailun Wang, Yanping Xia, Hongxin Gao, Keming Luo, Zheng Cao, Juan Qi
ABSTRACT
In this work, poly(ethylene terephthalate) (PET) chain-extending products with different molecular weights were prepared by reactive extrusion using isocyanate trimer (C-HK) as the trifunctional chain extender. The effect of the chain extender C-HK on the intrinsic viscosity, melt flow property, crystallization behavior, crystallization morphology, and mechanical property of PET was investigated. The results showed that when the content of the chain extender was increased from 0.6 to 1.4 wt%, the viscosity average molecular weight of PET was effectively increased from 2.36 × 104 to 5.46 × 104 g·mol-1. After the chain extending, the crystallinity and the time of semicrystallization of PET were significantly decreased. After the isothermal crystallization at 220 °C for 5 min, the spherulites formed by pure PET became larger. With the increase in molecular weight of PET after chain extension, its spherulite size was significantly decreased without changing the crystalline structure. The chain-extended PET also exhibited more excellent bending-resistant and impact-resistant properties. While the tensile strength of PET after chain extension was slightly decreased, the bending strength was increased by a maximum value of 56.8%, and the impact strength was increased by a maximum value of five times.
Sigma-Aldrich
Tetramethylammonium acetate, ≥97%