MilliporeSigma
  • Electron-Deficient Alkynes as Powerful Tools against Root-Knot Nematode Melodogyne incognita: Nematicidal Activity and Investigation on the Mode of Action.
Electron-Deficient Alkynes as Powerful Tools against Root-Knot Nematode Melodogyne incognita: Nematicidal Activity and Investigation on the Mode of Action.
Journal of agricultural and food chemistry (2020-09-15)
Graziella Tocco, Kodjo Eloh, Antonio Laus, Nicola Sasanelli, Pierluigi Caboni
ABSTRACT
The present study reports on the powerful nematicidal activity of a series of electron-deficient alkynes against the root-knot nematode Meloidogyne incognita (Kofoid and White) Chitwood. Interestingly, we found that the conjugation of electron-withdrawing carbonyl groups to an alkyne triple bond was extremely proficient in inducing nematode paralysis and death. In particular, dimethylacetylenedicarboxylate (10), 3-butyn-2-one (1), and methyl propiolate (4), with EC50/48 h of 1.54 ± 0.16, 2.38 ± 0.31, and 2.83 ± 0.28 mg/L, respectively, were shown to be the best tested compounds. Earlier studies reported on the ability of alkynoic esters and alkynones to induce a chemoselective cysteine modification of unprotected peptides. Thus, also following our previous findings on the impairment of vacuolar-type proton translocating ATPase functionality by activated carbonyl derivatives, we speculate that the formation of a vinyl sulfide linkage might be responsible for the nematicidal activity of the presented electron-deficient alkynes.
Sigma-Aldrich
Methyl propiolate, 99%