MilliporeSigma
  • Dstac Regulates Excitation-Contraction Coupling in Drosophila Body Wall Muscles.
Dstac Regulates Excitation-Contraction Coupling in Drosophila Body Wall Muscles.
Frontiers in physiology (2020-10-31)
I-Uen Hsu, Jeremy W Linsley, Lilly E Reid, Richard I Hume, Ari Leflein, John Y Kuwada
ABSTRACT
Stac3 regulates excitation-contraction coupling (EC coupling) in vertebrate skeletal muscles by regulating the L-type voltage-gated calcium channel (Cav channel). Recently a stac-like gene, Dstac, was identified in Drosophila and found to be expressed by both a subset of neurons and muscles. Here, we show that Dstac and Dmca1D, the Drosophila L-type Cav channel, are necessary for normal locomotion by larvae. Immunolabeling with specific antibodies against Dstac and Dmca1D found that Dstac and Dmca1D are expressed by larval body-wall muscles. Furthermore, Ca2+ imaging of muscles of Dstac and Dmca1D deficient larvae found that Dstac and Dmca1D are required for excitation-contraction coupling. Finally, Dstac appears to be required for normal expression levels of Dmca1D in body-wall muscles. These results suggest that Dstac regulates Dmca1D during EC coupling and thus muscle contraction.
Sigma-Aldrich
Anti-Chicken IgY (H+L), highly cross-adsorbed, CF 633 antibody produced in donkey, ~2 mg/mL, affinity isolated antibody