MilliporeSigma
  • The causal effect of obesity on prediabetes and insulin resistance reveals the important role of adipose tissue in insulin resistance.
The causal effect of obesity on prediabetes and insulin resistance reveals the important role of adipose tissue in insulin resistance.
PLoS genetics (2020-09-15)
Zong Miao, Marcus Alvarez, Arthur Ko, Yash Bhagat, Elior Rahmani, Brandon Jew, Sini Heinonen, Linda Liliana Muñoz-Hernandez, Miguel Herrera-Hernandez, Carlos Aguilar-Salinas, Teresa Tusie-Luna, Karen L Mohlke, Markku Laakso, Kirsi H Pietiläinen, Eran Halperin, Päivi Pajukanta
ABSTRACT
Reverse causality has made it difficult to establish the causal directions between obesity and prediabetes and obesity and insulin resistance. To disentangle whether obesity causally drives prediabetes and insulin resistance already in non-diabetic individuals, we utilized the UK Biobank and METSIM cohort to perform a Mendelian randomization (MR) analyses in the non-diabetic individuals. Our results suggest that both prediabetes and systemic insulin resistance are caused by obesity (p = 1.2×10-3 and p = 3.1×10-24). As obesity reflects the amount of body fat, we next studied how adipose tissue affects insulin resistance. We performed both bulk RNA-sequencing and single nucleus RNA sequencing on frozen human subcutaneous adipose biopsies to assess adipose cell-type heterogeneity and mitochondrial (MT) gene expression in insulin resistance. We discovered that the adipose MT gene expression and body fat percent are both independently associated with insulin resistance (p≤0.05 for each) when adjusting for the decomposed adipose cell-type proportions. Next, we showed that these 3 factors, adipose MT gene expression, body fat percent, and adipose cell types, explain a substantial amount (44.39%) of variance in insulin resistance and can be used to predict it (p≤2.64×10-5 in 3 independent human cohorts). In summary, we demonstrated that obesity is a strong determinant of both prediabetes and insulin resistance, and discovered that individuals' adipose cell-type composition, adipose MT gene expression, and body fat percent predict their insulin resistance, emphasizing the critical role of adipose tissue in systemic insulin resistance.
Product Description
Flowmi® Cell Strainers, porosity 40 μm, for 1000 uL Pipette Tips, pack of 50 ea
Pricing and availability is not currently available.