MilliporeSigma
  • Effect of intravitreal Rho kinase inhibitor ripasudil (K-115) on feline retinal microcirculation.
Effect of intravitreal Rho kinase inhibitor ripasudil (K-115) on feline retinal microcirculation.
Experimental eye research (2015-07-22)
Seigo Nakabayashi, Motofumi Kawai, Takafumi Yoshioka, Yong-Seok Song, Tomofumi Tani, Akitoshi Yoshida, Taiji Nagaoka
ABSTRACT
Ripasudil (K-115) is a novel Rho kinase inhibitor with a potent intraocular pressure-lowering effect. However, it is unclear whether ripasudil affects the retinal blood flow (RBF). We investigated the effect of ripasudil on feline retinal microcirculation. Ripasudil (5 μM, 50 μM or 5 mM; n = 5 each concentration) or vehicle (PBS; n = 5) was injected intravitreally. The vessel diameter (D) and blood velocity (V) were measured by laser Doppler velocimetry simultaneously in the first-order retinal arterioles and the RBF was calculated. The measurements started 5 min before the injection and were performed every 10 min for 120 min. After the intravitreal injection, the retinal circulatory parameters did not change significantly in PBS or 5 μM of ripasudil. The blood V and RBF increased significantly compared to baseline, whereas the vessel D did not change significantly in 50 μM and 5 mM of ripasudil. The V in 50 μM, and the V and RBF in 5 mM of ripasudil significantly increased compared to those in PBS. Intravitreal administration of ripasudil increased the blood V and RBF in cats, suggesting that ripasudil has the potential to improve the retinal blood flow.
Sigma-Aldrich
Ripasudil Hydrochloride, ≥98% (HPLC)