MilliporeSigma
  • WAY-163909 [(7bR, 10aR)-1,2,3,4,8,9,10,10a-octahydro-7bH-cyclopenta-[b][1,4]diazepino[6,7,1hi]indole], a novel 5-hydroxytryptamine 2C receptor-selective agonist with anorectic activity.
WAY-163909 [(7bR, 10aR)-1,2,3,4,8,9,10,10a-octahydro-7bH-cyclopenta-[b][1,4]diazepino[6,7,1hi]indole], a novel 5-hydroxytryptamine 2C receptor-selective agonist with anorectic activity.
The Journal of pharmacology and experimental therapeutics (2005-02-12)
John Dunlop, Annmarie L Sabb, Hossein Mazandarani, Jean Zhang, Sachin Kalgaonker, Eugenia Shukhina, Stacey Sukoff, Robert L Vogel, Gary Stack, Lee Schechter, Boyd L Harrison, Sharon Rosenzweig-Lipson
ABSTRACT
The pharmacological profile of WAY-163909 [(7bR, 10aR)-1,2,3,4,8,9,10,10a-octahydro-7bH-cyclopenta-[b][1,4]diazepino[6,7,1hi]indole], a novel 5-hydroxytryptamine (HT)(2C) (serotonin) receptor-selective agonist is presented. WAY-163909 displaced [(125)I]2,5-dimethoxy-4-iodoamphetamine binding from human 5-HT(2C) receptor sites, in Chinese hamster ovary (CHO) cell membranes, with a K(i) value of 10.5 +/- 1.1 nM. Binding affinities determined for the human 5-HT(2A) and 5-HT(2B) receptor subtypes were 212 and 485 nM, respectively. In functional studies, WAY-163909 stimulated the mobilization of intracellular calcium in CHO cells stably expressing the human 5-HT(2C) receptor with an EC(50) value of 8 nM, and E(max) relative to 5-HT of 90%. WAY-163909 failed to stimulate calcium mobilization in cells expressing the human 5-HT(2A) receptor subtype (EC(50) > 10muM) and was a 5-HT(2B) receptor partial agonist (EC(50) 185 nM, E(max) 40%). WAY-163909 exhibited negligible affinity (<50% inhibition at 1 muM) for other receptor sites examined, including human 5-HT(1A), D2, and D3 receptors, and the 5-HT transporter binding site in rat cortical membranes. WAY-163909 exhibited weak affinity for the human D4 (245 nM) and 5-HT(7) (343 nM) receptor subtypes and the alpha1 binding site in rat cortical membranes (665 nM). WAY-163909 produced a dose-dependent reduction in food intake in normal Sprague-Dawley rats (ED(50) = 2.93 mg/kg), an effect blocked by a 5-HT(2C) receptor antagonist but not by a 5-HT(2A) or 5-HT(2B) receptor antagonist. In addition, WAY-163909 decreased food intake in obese Zucker rats and diet-induced obese mice with ED(50) values of 1.4 and 5.19 mg/kg i.p., respectively, consistent with the potential utility of 5-HT(2C) receptor agonists as anti-obesity agents.
Sigma-Aldrich
WAY-163909 hydrochloride, ≥98% (HPLC)