MilliporeSigma
  • Generation of a human iPSC line CIBi007-A from a patient with young-onset Parkinson's disease carrying variants in PRKN and HTRA2.
Generation of a human iPSC line CIBi007-A from a patient with young-onset Parkinson's disease carrying variants in PRKN and HTRA2.
Stem cell research (2020-07-21)
Yubo Yan, Suihan Wu, Lixiang Jiang, Bo Yang, Xuhui Chen, Yuqing Liu, Baorong Yu, Jian Fu, Jin Yu, Hui Ning, Meng Chu, Jiayin Yang
ABSTRACT
Variations in PRKN or HTRA2 are associated with Parkinson's disease. We generated a human induced pluripotent stem cell (iPSC) line CIBi007-A from a patient with young-onset Parkinson's disease (YOPD) who carried variants in PRKN and HTRA2. The generated iPSCs resembled human embryonic stem cells, expressed pluripotency markers, exhibited a normal karyotype, and could be differentiated into three germ layers in vitro. This line will be valuable for investigating disease mechanisms of YOPD and screening candidate drugs.