MilliporeSigma
  • Epigallocatechin gallate and gallocatechin gallate in green tea catechins inhibit extracellular release of Vero toxin from enterohemorrhagic Escherichia coli O157:H7.
Epigallocatechin gallate and gallocatechin gallate in green tea catechins inhibit extracellular release of Vero toxin from enterohemorrhagic Escherichia coli O157:H7.
Biochimica et biophysica acta (1999-11-26)
Y Sugita-Konishi, Y Hara-Kudo, F Amano, T Okubo, N Aoi, M Iwaki, S Kumagai
ABSTRACT
We studied the effects of six catechin derivatives (catechin, epigallocatechin, epicatechin, epicatechin gallate, epigallocatechin gallate (EGCg) and gallocatechin gallate (GCg)) in green tea on the production and extracellular release of Vero toxins (VTs) from enterohemorrhagic Escherichia coli (EHEC) cultured at 37 degrees C for 24 h. EGCg and GCg in the culture medium markedly inhibited extracellular VTs release from EHEC cells into the culture supernatant fluid at concentrations of 0.05 mg/ml or higher, as estimated by both the reversed passive latex agglutination assay and cytotoxic assay using Vero cells. Production and extracellular release of maltose binding protein, a periplasmic protein, into the culture supernatant were also inhibited by EGCg and GCg, indicating that their inhibitory effect on release from periplasm into the outer milieu is not specific to VTs, but general to the proteins accumulated in EHEC periplasm.
Sigma-Aldrich
(−)-Gallocatechin gallate, from green tea, ≥98% (HPLC)