MilliporeSigma
  • Liver receptor homolog-1 (NR5a2) regulates CD95/Fas ligand transcription and associated T-cell effector functions.
Liver receptor homolog-1 (NR5a2) regulates CD95/Fas ligand transcription and associated T-cell effector functions.
Cell death & disease (2017-04-14)
Juliane Schwaderer, Ann-Kathrin Gaiser, Truong San Phan, M Eugenia Delgado, Thomas Brunner
ABSTRACT
CD95/Fas ligand (FasL) is a cell death-promoting member of the tumor necrosis factor family with important functions in the regulation of T-cell homeostasis and cytotoxicity. In T cells, FasL expression is tightly regulated on a transcriptional level involving a complex set of different transcription factors. The orphan nuclear receptor liver receptor homolog-1 (LRH-1/NR5a2) is involved in the regulation of development, lipid metabolism and proliferation and is predominantly expressed in epithelial tissues. However, its expression in T lymphocytes has never been reported so far. Based on in silico analysis, we identified potential LRH-1 binding sites within the FASLG promoter. Here, we report that LRH-1 is expressed in primary and secondary lymphatic tissues, as well as in CD4+ and CD8+ T cells. LRH-1 directly binds to its binding sites in the FASLG promoter, and thereby drives FASLG promoter activity. Mutations in the LRH-1 binding sites reduce FASLG promoter activity. Pharmacological inhibition of LRH-1 decreases activation-induced FasL mRNA expression, as well as FasL-mediated activation-induced T-cell apoptosis and T-cell cytotoxicity. In a mouse model of Concanavalin A-induced and FasL-mediated hepatitis pharmacological inhibition of LRH-1 resulted in decreased hepatic FasL expression and a significant reduction of liver damage. In summary, these data show for the first time LRH-1 expression in T cells, its role in FASLG transcription and the potential of pharmacological inhibition of LRH-1 in the treatment of FasL-mediated immunopathologies.
Sigma-Aldrich
SR1848, ≥98% (HPLC)