MilliporeSigma
  • Macroautophagy is impaired in old murine brain tissue as well as in senescent human fibroblasts.
Macroautophagy is impaired in old murine brain tissue as well as in senescent human fibroblasts.
Redox biology (2016-11-09)
Christiane Ott, Jeannette König, Annika Höhn, Tobias Jung, Tilman Grune
ABSTRACT
The overall decrease in proteolytic activity in aging can promote and accelerate protein accumulation and metabolic disturbances. To specifically analyze changes in macroautophagy (MA) we quantified different autophagy-related proteins (ATGs) in young, adult and old murine tissue as well as in young and senescent human fibroblasts. Thus, we revealed significantly reduced levels of ATG5-ATG12, LC3-II/LC3-I ratio, Beclin-1 and p62 in old brain tissue and senescent human fibroblasts. To investigate the role of mTOR, the protein itself and its target proteins p70S6 kinase and 4E-BP1 were quantified. Significant increased mTOR protein levels were determined in old tissue and cells. Determination of phosphorylated and basal amount of both proteins suggested higher mTOR activity in old murine tissue and senescent human fibroblasts. Besides the reduced levels of ATGs, mTOR can additionally reduce MA, promoting further acceleration of protein accumulation and metabolic disturbances during aging.
Sigma-Aldrich
Concanamycin A, ≥70% (HPLC)
Pricing and availability is not currently available.