MilliporeSigma
  • Identification and characterization of an interleukin-15 homologue from Tetraodon nigroviridis.
Identification and characterization of an interleukin-15 homologue from Tetraodon nigroviridis.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2006-02-04)
Wei Fang, Li-Xin Xiang, Jian-Zhong Shao, Yi Wen, Shu-Yun Chen
ABSTRACT
Interleukin-15 (IL-15) plays an important role in adaptive immune systems in vertebrates with similar bioactivities to interleukin-2 (IL-2). Here we report molecular cloning, sequence analysis and distribution of an IL-15 homologue from a pufferfish (Tetraodon nigroviridis). It is located within a 3,088 bp genomic fragment, transcribed into a 1,056 bp mRNA including 158 bp 5'UTR (untranslated region), 519 bp ORF (open reading frame) and 379 bp 3'UTR. T. nigroviridis IL-15 is constitutively detectable in tissues and organs selected. Levels of transcripts were observed after various stimulations. Gene organization is similar to mammals and birds, and a high degree of conservation of chromosome synteny exists between them. Systematic genomics search against Takifugu rubripes genome supports our conclusions. The T. nigroviridis IL-15 precursor with 172aa (amino acids) contains a putative 53aa signal peptide, while the mature peptide has a calculated molecular mass of 13.36 kDa and a theoretical pI of 4.67. The protein sequence shares 13.3-62.1% identity with reported IL-15s. Phylogenetic analysis grouped Tetraodon with other fish on a separated branch, excluded from mammalian and avian IL-15s. In addition, our analysis on another annotated T. nigroviridis IL-15 demonstrated that it may be a paralogue of IL-15. To differentiate it from the known IL-15s, we described it as IL-15x.
Sigma-Aldrich
Human IL-15 ELISA Kit, for serum, plasma, cell culture supernatant and urine