MilliporeSigma
  • Graphene Surfaces Interaction with Proteins, Bacteria, Mammalian Cells, and Blood Constituents: The Impact of Graphene Platelet Oxidation and Thickness.
Graphene Surfaces Interaction with Proteins, Bacteria, Mammalian Cells, and Blood Constituents: The Impact of Graphene Platelet Oxidation and Thickness.
ACS applied materials & interfaces (2020-04-03)
Patrícia C Henriques, Andreia T Pereira, Ana L Pires, André M Pereira, Fernão D Magalhães, Inês C Gonçalves
ABSTRACT
Graphene-based materials (GBMs) have been increasingly explored for biomedical applications. However, interaction between GBMs-integrating surfaces and bacteria, mammalian cells, and blood components, that is, the major biological systems in our body, is still poorly understood. In this study, we systematically explore the features of GBMs that most strongly impact the interactions of GBMs films with plasma proteins and biological systems. Films produced by vacuum filtration of GBMs with different oxidation degree and thickness depict different surface features: graphene oxide (GO) and few-layer GO (FLGO) films are more oxidized, smoother, and hydrophilic, while reduced GO (rGO) and few-layer graphene (FLG) are less or nonoxidized, rougher, and more hydrophobic. All films promote glutathione oxidation, although in a lower extent by rGO, indicating their potential to induce oxidative stress in biological systems. Human plasma proteins, which mediate most of the biological interactions, adsorb less to oxidized films than to rGO and FLG. Similarly, clinically relevant bacteria, Staphylococcus epidermidis, Staphylococcus aureus, Pseudomonas aeruginosa, and Escherichia coli, adhere less to GO and FLGO films, while rGO and FLG favor bacterial adhesion and viability. Surface features caused by the oxidation degree and thickness of the GBMs powders within the films have less influence toward human foreskin fibroblasts; all materials allow cell adhesion, proliferation and viability up to 14 days, despite less on rGO surfaces. Blood cells adhere to all films, with higher numbers in less or nonoxidized surfaces, despite none having caused hemolysis (<5%). Unlike thickness, oxidation degree of GBMs platelets strongly impact surface morphology/topography/chemistry of the films, consequently affecting protein adsorption and thus bacteria, fibroblasts and blood cells response. Overall, this study provides useful guidelines regarding the choice of the GBMs to use in the development of surfaces for an envisioned application. Oxidized materials appear as the most promising for biomedical applications that require low bacterial adhesion without being cytotoxic to mammalian cells.
Product Description
Millipore
Nylon Membrane Filter, 20.0 μm Pore Size, Millipore, filter diam. 47 mm, hydrophilic