MilliporeSigma
  • Evaluation of the exposure, dose-response and fate in the lung and pleura of chrysotile-containing brake dust compared to TiO2, chrysotile, crocidolite or amosite asbestos in a 90-day quantitative inhalation toxicology study - Interim results Part 1: Experimental design, aerosol exposure, lung burdens and BAL.
Evaluation of the exposure, dose-response and fate in the lung and pleura of chrysotile-containing brake dust compared to TiO2, chrysotile, crocidolite or amosite asbestos in a 90-day quantitative inhalation toxicology study - Interim results Part 1: Experimental design, aerosol exposure, lung burdens and BAL.
Toxicology and applied pharmacology (2019-12-15)
D M Bernstein, B Toth, R A Rogers, D E Kling, P Kunzendorf, J I Phillips, H Ernst
ABSTRACT
This 90-day repeated-dose inhalation toxicology study of brake-dust (BD) (brakes manufactured with chrysotile) in rats provides a comprehensive understanding of the biokinetics and potential toxicology in the lung and pleura. Exposure was 6 h/d, 5d/wk., 13wks followed by lifetime observation (~20 % survival). Control groups included a particle control (TiO2), chrysotile, commercial crocidolite and amosite asbestos. Aerosol fiber distributions of the chrysotile, crocidolite and amosite were similar (fibers L > 20 μm/cm3: chrysotile-Low/High 29/72; crocidolite 24; amosite 47 fibers/cm3; WHO-fibers/cm3: chrysotile-Low/High 119/233; crocidolite 181; amosite 281 fibers/cm3). The number of particles/cm3 in the BD was similar to that in the chrysotile, crocidolite & amosite exposures (BD 470-715; chrysotile 495-614; crocidolite 415; amosite 417 particles/cm3). In the BD groups, few fibers L > 20 μm were observed in the lungs at the end of exposure and no fibers L > 20 μm at 90d post exposure. In the chrysotile groups, means of 204,000 and 290,000 fibers(L > 20 μm)/lung were measured at 89d. By 180d, means of 1 and 3.9 fibers were counted on the filter corresponding to 14,000 and 55,000 fibers(L > 20 μm)/lung. In the crocidolite and amosite groups mean lung concentrations were 9,055,000 and 11,645,000 fibers(L > 20 μm)/lung at 89d. At 180d the means remained similar with 8,026,000 and 11,591,000 fibers(L > 20 μm)/lung representing 10-13% of the total lung fibers. BAL determined the total number of macrophages, lymphocytes, neutrophils, eosinophils, epithelial-cells and IL-1 beta, TNF-alpha and TGF-beta. At the moderate aerosol concentrations used in this study, neutrophil counts increased ~5 fold in the amphibole asbestos exposure groups. All other groups and parameters showed no important differences at these exposure concentrations. The exposure and lung burden results provide a sound basis for assessing the potential toxicity of the brake dust in comparison to the TiO2 particle control and the chrysotile, crocidolite and amosite asbestos control groups. The BAL results provide an initial indication of the differential response. Part 2 presents the presentation and discussion of the histopathological and confocal microscopy findings in this study through 90 days post exposure.
Product Description
Millipore
PVDF Membrane Filter, 0.45 μm Pore Size, Durapore®, filter diam. 47 mm, hydrophilic, white
Pricing and availability is not currently available.