MilliporeSigma
  • First in-human PET study and kinetic evaluation of [18F]AS2471907 for imaging 11β-hydroxysteroid dehydrogenase type 1.
First in-human PET study and kinetic evaluation of [18F]AS2471907 for imaging 11β-hydroxysteroid dehydrogenase type 1.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2019-03-22)
Shivani Bhatt, Nabeel B Nabulsi, Songye Li, Zhengxin Cai, David Matuskey, Jason Bini, Soheila Najafzadeh, Michael Kapinos, Jim R Ropchan, Richard E Carson, Kelly P Cosgrove, Yiyun Huang, Ansel T Hillmer
ABSTRACT
11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) catalyzes enzymatic conversion of cortisone into the stress hormone cortisol. This first-in-human brain imaging study characterizes the kinetic modeling and test-retest reproducibility of [18F]AS2471907, a novel PET radiotracer for 11β-HSD1. Eight individuals underwent one 180-min (n = 4) or two 240-min (n = 4) [18F]AS2471907 PET brain scans (12 total) acquired on the high-resolution research tomograph (HRRT) scanner with arterial blood sampling. Imaging data were modeled with 1-tissue (1T) and 2-tissue (2T) compartment models and with multilinear analysis (MA1) to estimate [18F]AS2471907 availability (VT). [18F]AS2471907 demonstrated high, heterogeneous uptake throughout the brain. Of the compartment models, 2T best described [18F]AS2471907 data. Estimates of VT were highly correlated between 2T and MA1 (t* = 30 min) with MA1 yielding VT values ranging from 3.2 ± 1.0 mL/cm3 in the caudate to 15.7 ± 4.2 mL/cm3 in the occipital cortex. The median absolute test-retest variability of 16 ± 5% and high intraclass correlation coefficient (ICC) values of 0.67-0.97 across regions indicate fair test-retest reliability but large intersubject variability. VT estimates using 180 min were within 10% of estimates using full acquisition time. In summary, [18F]AS2471907 exhibits reasonable kinetic properties for imaging 11β-HSD1 in human brain.
Product Description
Millipore
Centrifree® Ultrafiltration Centrifugal Filters, MWCO 30 kDa, sample volume 1 mL, Ultracel® PL regenerated cellulose membrane