MilliporeSigma
  • Suberoyl bis-hydroxamic acid activates Notch-1 signaling and induces apoptosis in medullary thyroid carcinoma cells.
Suberoyl bis-hydroxamic acid activates Notch-1 signaling and induces apoptosis in medullary thyroid carcinoma cells.
The oncologist (2008-02-29)
Li Ning, David Yu Greenblatt, Muthusamy Kunnimalaiyaan, Herbert Chen
ABSTRACT
Medullary thyroid carcinoma (MTC) is a neuroendocrine (NE) malignancy that frequently metastasizes and has limited treatments. We recently reported that ectopic expression of Notch-1 in human MTC cells suppresses growth. The objective of this study was to evaluate the ability of suberoyl bis-hydroxamic acid (SBHA) to modulate Notch-1 signaling in MTC cells. At baseline, no active Notch-1 protein was present in MTC cells. Treatment with SBHA resulted in a dose-dependent induction of the Notch-1 intracellular domain, the active form of the protein. Furthermore, with Notch-1 activation there was a concomitant decrease in achaete-scute complex-like 1 (ASCL-1), a downstream target of Notch-1 signaling, as well as the NE tumor marker chromogranin A (CgA). Transfection of Notch-1 small-interfering RNA into MTC cells blocked the effects of SBHA on Notch-1 activation, ASCL-1, and CgA. Importantly, SBHA treatment resulted in a dose-dependent decrease in cell viability. Treated cells had an increase in protein levels of cleaved caspase-3 and poly ADP-ribose polymerase, and changes in the expression of apoptotic mediators including Bcl-X(L) and Bad, indicating that the growth inhibition was a result of apoptosis. These results demonstrate that SBHA activates Notch-1 signaling, which is associated with the antiproliferative and apoptotic effects in MTC cells. Therefore, Notch-1 activation with SBHA is an attractive new strategy for the treatment of patients with MTC.
Sigma-Aldrich
Suberohydroxamic acid, 95%
Pricing and availability is not currently available.