MilliporeSigma
  • Leucine-enkephalin-immunoreactive neurons in the brain of the cichlid fish Oreochromis mossambicus.
Leucine-enkephalin-immunoreactive neurons in the brain of the cichlid fish Oreochromis mossambicus.
Neuropeptides (2019-12-18)
Vijayalaxmi, Amul J Sakharkar, C B Ganesh
ABSTRACT
Enkephalins are the pentapeptides involved in pain relief and neuroendocrine responses with high affinity for delta opioid receptors in vertebrates. In the present investigation, we studied the distribution of leucine-enkephalin-immunoreactive (L-ENK-ir) neurons in the brain of the cichlid fish Oreochromis mossambicus. Application of the antisera against L-ENK revealed the presence of numerous L-ENK-ir perikarya and fibres in subdivisions of the dorsal and the ventral telencephalon, the medial olfactory tract and the nucleus entopeduncularis, whereas intensely labelled L-ENK-ir fibres were noticed in the olfactory bulb. Furthermore, the presence of L-ENK-ir cells and dense accumulations of fibres in the preoptic area and its subdivisions, the nucleus preopticus pars magnocellularis and the nucleus preopticus pars parvocellularis suggested a role for this peptide in regulation of reproduction. While intensely labelled cells and fibres were found in the nucleus lateralis tuberis pars lateralis as well as the nucleus lateralis tuberis pars medialis, some L-ENK-ir fibres were seen at the hypothalamo-hypophyseal tract indicating the possible hypophysiotrophic role for this peptide. Numerous L-ENK-ir cells and dense network of fibres were observed in the subdivisions of the nucleus recess lateralis and the pretectal area, whereas intensely labelled thick network of L-ENK- fibres were found in the ventromedial thalamic nucleus, the sub-layers of the optic tectum and the rostral spinal cord. The widespread distribution of L-ENK-immunoreactivity in the olfactory bulb, the telencephalon, the diencephalon and the mesencephalon regions of the brain as well as the spinal cord suggests the possible involvement of this peptide in the regulation of diverse functions such as neuroendocrine, antinociceptive, visual and olfactory responses in O. mossambicus.