MilliporeSigma
  • Palladium-catalyzed regio- and stereoselective synthesis of aryl and 3-indolyl-substituted 4-methylene-3,4-dihydroisoquinolin-1(2H)-ones.
Palladium-catalyzed regio- and stereoselective synthesis of aryl and 3-indolyl-substituted 4-methylene-3,4-dihydroisoquinolin-1(2H)-ones.
Beilstein journal of organic chemistry (2020-06-20)
Valeria Nori, Antonio Arcadi, Armando Carlone, Fabio Marinelli, Marco Chiarini
ABSTRACT
Cascade cyclocarbopalladation of the readily available aryl/alkyl-substituted propargylic amides containing an aryl iodide moiety, followed by Suzuki-Miyaura coupling with arylboronic acids, allowed an efficient regio- and stereoselective synthesis of tetrasubstituted 4-methylene-3,4-dihydroisoquinolin-1(2H)-ones. Moreover, cascade cyclocarbopalladation, followed by the reaction with 2-alkynyltrifluoroacetanilides, accomplished a double cyclization to afford challenging 4-methylene-3,4-dihydroisoquinolin-1(2H)-ones bearing a 3-indolyl substituent through aminopalladation/reductive elimination.
Sigma-Aldrich
3-Hydroxycoumarin
Pricing and availability is not currently available.