MilliporeSigma
  • Epigenetic activation of O-linked β-N-acetylglucosamine transferase overrides the differentiation blockage in acute leukemia.
Epigenetic activation of O-linked β-N-acetylglucosamine transferase overrides the differentiation blockage in acute leukemia.
EBioMedicine (2020-04-10)
K M Kampa-Schittenhelm, T Haverkamp, M Bonin, V Tsintari, H J Bühring, L Haeusser, G Blumenstock, S T Dreher, T Ganief, F Akmut, B Illing, U A Mau-Holzmann, I Bonzheim, E Schleicher, W Vogel, M M Schittenhelm
ABSTRACT
Overriding the differentiation blockage in acute myeloid leukemia (AML) is the most successful mode-of-action in leukemia therapy - now curing the vast majority of patients with acute promyelocytic leukemia (APL) using all-trans retinoic acid (ATRA)-based regimens. Similar approaches in other leukemia subtypes, such as IDH1/2-mutated AML, are under active investigation. We herein present successful release of the differentiation blockage upon treatment with the natural (-)-Δ9-Tetrahydrocannabinol isomer dronabinol in vitro and in vivo. Cellular maturation and differentiation were followed in two patients employing whole genome methylation profiling, proteome analyses, NGS deep sequencing and multispectral imaging flow cytometry. For functional studies lentiviral OGT knock-down in vitro and ex vivo cell models were created to evaluate proliferative, apoptotic and differentiating effects of OGT in acute leukemia. In here, we provide molecular evidence that dronbinol is capable to override the differentiation blockage of acute leukemia blasts at the state of the leukemia-initiating clone. We further identify the O-linked β-N-acetyl glucosamine (O-GlcNAc) transferase (OGT) to be crucial in this process. OGT is a master regulator enzyme adding O-GlcNAc to serine or threonine residues in a multitude of target proteins. Aberrant O-GlcNAc modification is implicated in pathologies of metabolic, neurodegenerative and autoimme diseases as well as cancers. We provide evidence that dronabinol induces transcription of OGT via epigenetic hypomethylation of the transcription start site (TSS). A lentiviral OGT-knock out approach proves the central role of OGT exerting antileukemic efficacy via a dual-mechanism of action: High concentrations of dronabinol result in induction of apoptosis, whereas lower concentrations drive cellular maturation. Most intriguingly, overriding of the differentiation blockage of acute leukemia blasts is validated in vivo following two patients treated with dronabinol. In conclusion, we provide evidence for overcoming the differentiation blockage in acute leukemia in subentities beyond promyelocytic and IDH1/2-mutated leukemia and thereby identify O-GlcNAcylation as a novel (drugable) field for future leukemia research. Unrestricted grant support by the IZKF Program of the Medical Faculty Tübingen (MMS) and Brigitte Schlieben-Lange Program as well as the Margarete von Wrangell Program of the Ministry of Science, Research and the Arts, Baden-Württemberg, Germany (KKS) and Athene Program of the excellence initiative University of Tübingen (KKS).
Sigma-Aldrich
LY 320135, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human OGT