MilliporeSigma
  • Functional characterization of a binding protein for Type-II sex pheromones in the tea geometrid moth Ectropis obliqua Prout.
Functional characterization of a binding protein for Type-II sex pheromones in the tea geometrid moth Ectropis obliqua Prout.
Pesticide biochemistry and physiology (2020-05-04)
Yuting Yan, Yuxing Zhang, Xiaohui Tu, Qian Wang, Yujie Li, Hongyue Li, Qi Wang, Yongjun Zhang, Liang Sun
ABSTRACT
The tea geometrid moth Ectropis obliqua Prout is one of the most serious moth pests in tea plants, and its sex pheromones have been identified as typical Type-II polyunsaturated hydrocarbons and epoxide derivatives. Therefore, the E. obliqua male olfactory system provides a good model to study the molecular basis of Type-II sex pheromone recognition as well as functional gene evolution towards structurally different types of moth sex pheromones. In this study, we identified the full-length sequence of a pheromone-binding protein, EoblPBP2 and revealed that it clustered together with the lepidopteran PBP2 subfamily, which binds Type I acetate pheromones. These findings suggest that the EoblPBP2 sequence and physiological function are conserved, although E. obliqua evolved Type II hydrocarbon and epoxide sex pheromones structurally different from Type I acetates. To examine this hypothesis, we studied the expression patterns and in vitro functions of EoblPBP2 in detail. Quantitative real-time PCR experiments showed that EoblPBP2 was predominantly expressed in male E. obliqua antennae. Fluorescence in situ hybridization further demonstrated that the EoblPBP2 gene was abundantly expressed in the pheromone-sensitive sensilla trichodea Str-I in male E. obliqua. The physiological function of recombinant EoblPBP2 was then examined using a competitive binding assay. The results showed that EoblPBP2 had high affinities for three E. obliqua Type II sex pheromone components and Type I acetate pheromones in comparison to some plant volatiles. These results indicate that PBP2 is involved in the detection of Type II pheromones in E. obliqua and it still retains high binding affinities to acetate pheromones and some green leaf ester volatiles.