MilliporeSigma
  • Maternal 25-Hydroxyvitamin D Deficiency Promoted Metabolic Syndrome and Downregulated Nrf2/CBR1 Pathway in Offspring.
Maternal 25-Hydroxyvitamin D Deficiency Promoted Metabolic Syndrome and Downregulated Nrf2/CBR1 Pathway in Offspring.
Frontiers in pharmacology (2020-03-19)
Jianqiong Zheng, Xiaohui Liu, Bingbing Zheng, Zhenzhen Zheng, Hongping Zhang, Jiayong Zheng, Congcong Sun, Haiying Chen, Jie Yang, Zuo Wang, Meimei Lin, Jingjing Chen, Qingdiao Zhou, Zhi Zheng, Xiaoming Xu, Hao Ying
ABSTRACT
Metabolic syndrome is a disorder of energy use and storage, which is characterized by central obesity, dyslipidemia, and raised blood pressure and blood sugar levels. Maternal 25-hydroxyvitamin D deficiency is known to cause metabolic changes, chronic disease, and increased adiposity in adulthood. However, the underlying mechanism of induced metabolic syndrome (MetS) in the offspring in vitamin D deficient pregnant mothers remains unclear. We identified that maternal 25-hydroxyvitamin D deficiency enhances oxidative stress, which leads to the development of MetS in the mother and her offspring. Further, immunohistochemical, Western blotting, and qRT-PCR analyses revealed that maternal 25-hydroxyvitamin D deficiency inhibited the activation of the Nrf2/carbonyl reductase 1 (CBR1) pathway in maternal placenta, liver, and pancreas, as well as the offspring's liver and pancreas. Further analyses uncovered that application of 25-hydroxyvitamin D activated the Nrf2/CBR1 pathway, relieving the oxidative stress in BRL cells, suggesting that 25-hydroxyvitamin D regulates oxidative stress in offspring and induces the activation of the Nrf2/CBR1 pathway. Taken together, our study finds that maternal 25-hydroxyvitamin D deficiency is likely to result in offspring's MetS probably via abnormal nutrition transformation across placenta. Depression of the Nrf2/CBR1 pathway in both mothers and their offspring is one of the causes of oxidative stress leading to MetS. This study suggests that 25-hydroxyvitamin D treatment may relieve the offspring's MetS.
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3 (6,19,19-d3) solution, 50 μg/mL in ethanol, 97 atom % D, 96% (CP)
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human CBR1
Pricing and availability is not currently available.