MilliporeSigma
  • Biocontrol of Rice Seedling Rot Disease Caused by Curvularia lunata and Helminthosporium oryzae by Epiphytic Yeasts from Plant Leaves.
Biocontrol of Rice Seedling Rot Disease Caused by Curvularia lunata and Helminthosporium oryzae by Epiphytic Yeasts from Plant Leaves.
Microorganisms (2020-05-06)
Savitree Limtong, Parichat Into, Panchapond Attarat
ABSTRACT
Seedling rot disease in rice leads to significant loss in the production of seedlings. This research was conducted to explore yeasts that could be used as biological control agents against rice seedling rot disease caused by Curvularia lunata and Helminthosporium oryzae. In total, 167 epiphytic yeast strains were evaluated, revealing that 13 of these yeast strains demonstrated antagonistic activities against fungal pathogens and either C. lunata DOAC 2313 or H. oryzae DOAC 2293. The volatile organic compounds (VOCs) and biofilm produced were possible antagonistic mechanisms in vitro for all the antagonistic yeast strains. Using nursery trays in a greenhouse, this study evaluated the control of rice seedling rot disease caused by these two fungal pathogens using antagonistic yeasts, identified in the present study and from our previous study. Torulaspora indica DMKU-RP31 and Wickerhamomyces anomalus YE-42 were found to completely control rice seedling rot disease caused by both of these fungal pathogens. Furthermore, W. anomalus DMKU-RP04 revealed 100% disease control when the disease was caused by H. oryzae. This is the first report on using antagonistic yeasts to control rice seedling rot disease caused by C. lunata and H. oryzae. These three antagonistic yeasts also showed promising potential for development as biocontrol agents against rice seedling rot disease caused by fungi.
Sigma-Aldrich
Chromeazurol S, Dye content 45 %