MilliporeSigma
  • Aquaporin 1 affects pregnancy outcome and regulates aquaporin 8 and 9 expressions in the placenta.
Aquaporin 1 affects pregnancy outcome and regulates aquaporin 8 and 9 expressions in the placenta.
Cell and tissue research (2020-06-17)
Hui Luo, Yi Liu, Yizuo Song, Ying Hua, Xueqiong Zhu
ABSTRACT
To explore the effects of aquaporin (AQP) 1 on pregnancy outcome and the association between expression of AQP1 and other AQPs in the placenta and foetal membranes, the rate of copulatory plugs and pregnancy, amniotic fluid (AF) volume, osmolality and composition were determined in AQP1-knockout (AQP1-/-) mice at different gestational days (GD). The expression and location of AQP1 and other AQPs in the placenta and foetal membranes of AQP1-/- mice, AQP1-siRNA transfected WISH cells and oligohydramnios patients were also detected. Compared to control mice, AQP1-/- mice exhibited reduced copulation plug and successful pregnancy rates, but these effects were accompanied by a larger AF volume and lower AF osmolality at late gestation. AQP9 expression was significantly decreased in the placenta and foetal membranes of AQP1-/- mice, while AQP8 level was elevated in the foetal membranes of AQP1-/- mice. Moreover, AQP9 expression was suppressed in WISH cells after AQP1 downregulation. Furthermore, AQP9 expression was associated with AQP1 level in the placenta and foetal membranes in oligohydramnios. AQP1 may play a critical role in regulating pregnancy outcome and maternal-foetal fluid homeostasis. Changes in AQP1 expression may lead to compensatory alterations in AQP8 and AQP9 expression in the placenta.
Sigma-Aldrich
MISSION® esiRNA, targeting human AQP1