MilliporeSigma
  • miR-499 promotes immature porcine Sertoli cell growth by the PI3K/AKT pathway by targeting the PTEN gene.
miR-499 promotes immature porcine Sertoli cell growth by the PI3K/AKT pathway by targeting the PTEN gene.
Reproduction (Cambridge, England) (2019-11-23)
Hu Gao, Maoliang Ran, Hui Luo, Bo Weng, Xiangwei Tang, Yao Chen, An'qi Yang, Bin Chen
ABSTRACT
Sertoli cells are indispensable for normal spermatogenesis and increasing evidence has shown that microRNAs (miRNAs) participate in the regulation of Sertoli cell growth. However, the functions and regulatory mechanisms of miRNAs in Sertoli cells of domestic animals have not been fully investigated. In the present study, we mainly investigated the regulatory roles of miR-499 in immature porcine Sertoli cells. The results showed that miR-499 was mainly located in the basement section of seminiferous tubules of prepubertal porcine testicular tissue. Overexpression of miR-499 promoted cell proliferation and inhibited apoptosis, whereas miR-499 inhibition resulted in the opposite effects. The PTEN gene was directly targeted by miR-499, and the expression of mRNA and protein was also negatively regulated by miR-499 in immature porcine Sertoli cells. siRNA-induced PTEN knockdown resulted in a similar effect as overexpression of miR-499, and abolished the effects of miR-499 inhibition on immature porcine Sertoli cells. Moreover, both miR-499 overexpression and the PTEN knockdown activated the PI3K/AKT signaling pathway, whereas inhibition of the PI3K/AKT signaling pathway caused immature porcine Sertoli cell apoptosis and inhibited cell proliferation. Overall, miR-499 promotes proliferation and inhibits apoptosis in immature porcine Sertoli cells through the PI3K/AKT pathway by targeting the PTEN gene. This study provides novel insights into the effects of miR-499 in spermatogenesis through the regulation of immature Sertoli cell proliferation and apoptosis.
Sigma-Aldrich
MISSION® esiRNA, targeting human PTEN