MilliporeSigma
  • Ion-Mobility-Based Liquid Chromatography-Mass Spectrometry Quantitation of Taste-Enhancing Octadecadien-12-ynoic Acids in Mushrooms.
Ion-Mobility-Based Liquid Chromatography-Mass Spectrometry Quantitation of Taste-Enhancing Octadecadien-12-ynoic Acids in Mushrooms.
Journal of agricultural and food chemistry (2020-04-28)
Verena Karolin Mittermeier, Katja Pauly, Andreas Dunkel, Thomas Hofmann
ABSTRACT
For the accurate quantitation of kokumi-enhancing and bitter-tasting octadecadien-12-ynoic and octadecadienoic acids in chanterelles (Cantharellus cibarius Fr.), a sensitive ultra-high-performance liquid chromatography-differential ion mobility spectrometry-tandem mass spectrometry method was developed. On the basis of these quantitative data and the taste thresholds, dose-over-threshold factors were calculated to determine the contribution of these sensometabolites to the kokumi and bitter taste of chanterelles; e.g., 14,15-dehydrocrepenynic acid (3) and (9Z,15E)-14-oxooctadeca-9,15-dien-12-ynoic acid (7) were identified as key kokumi substances in raw chanterelles. Quantitative profiling of these compounds in various mushroom species demonstrated a unique accumulation of octadecadien-12-ynoic acids in Cantharellus. Furthermore, storage experiments highlighted dynamic processes, including the biosynthesis of these substances as a result of lipid peroxidation mechanisms.