MilliporeSigma
  • In vivo silencing of miR-125a-3p promotes myelin repair in models of white matter demyelination.
In vivo silencing of miR-125a-3p promotes myelin repair in models of white matter demyelination.
Glia (2020-03-13)
Davide Marangon, Enrica Boda, Roberta Parolisi, Camilla Negri, Corinna Giorgi, Francesca Montarolo, Simona Perga, Antonio Bertolotto, Annalisa Buffo, Maria P Abbracchio, Davide Lecca
ABSTRACT
In the last decade, microRNAs have been increasingly recognized as key modulators of glial development. Recently, we identified miR-125a-3p as a new player in oligodendrocyte physiology, regulating in vitro differentiation of oligodendrocyte precursor cells (OPCs). Here, we show that miR-125a-3p is upregulated in active lesions of multiple sclerosis (MS) patients and in OPCs isolated from the spinal cord of chronic experimental autoimmune encephalomyelitis (EAE) mice, but not in those isolated from the spontaneously remyelinating corpus callosum of lysolecithin-treated mice. To test whether a sustained expression of miR-125a-3p in OPCs contribute to defective remyelination, we modulated miR-125a-3p expression in vivo and ex vivo after lysolecithin-induced demyelination. We found that lentiviral over-expression of miR-125a-3p impaired OPC maturation, whereas its downregulation accelerated remyelination. Transcriptome analysis and luciferase reporter assay revealed that these effects are partly mediated by the direct interaction of miR-125a-3p with Slc8a3, a sodium-calcium membrane transporter, and identified novel candidate targets, such as Gas7, that we demonstrated necessary to correctly address oligodendrocytes to terminal maturation. These findings show that miR-125a-3p upregulation negatively affects OPC maturation in vivo, suggest its role in the pathogenesis of demyelinating diseases and unveil new targets for future promyelinating protective interventions.
Sigma-Aldrich
Indole-3-acetaldehyde–sodium bisulfite addition compound
Pricing and availability is not currently available.