Merck
  • Suppressed nuclear factor-kappa B alleviates lipopolysaccharide-induced acute lung injury through downregulation of CXCR4 mediated by microRNA-194.
Suppressed nuclear factor-kappa B alleviates lipopolysaccharide-induced acute lung injury through downregulation of CXCR4 mediated by microRNA-194.
Respiratory research (2020-06-12)
Ruidong Chen, Fei Xie, Jie Zhao, Bin Yue
ABSTRACT
Acute lung injury (ALI) is a highly lethal pulmonary disease that causes edema, hypoxemia and respiratory failure. Recent evidence indicates that nuclear factor-kappa B (NF-κB) plays a crucial role in ALI development. However, the regulatory mechanism of NF-κB on ALI remains enigmatic. In this study, we investigated potential molecular mechanism of NF-κB on ALI induced by lipopolysaccharide (LPS). BALB/c mice were subjected to intratracheal spraying of LPS to generate an ALI mode, with the activity of NF-κB in mice tissues being detected by enzyme linked immunosorbent assay (ELISA), and the number of inflammatory cells in bronchoalveolar lavage fluid being counted. Then, the macrophage cell line RAW264.7 exposed to LPS were treated with ammonium pyrrolidinedithiocarbamate (PDTC) (inhibitor of NF-κB), miR-194 mimic, or oe-chemokine receptor type 4 (CXCR4) separately or in combination. After that, ELISA and reverse transcription quantitative polymerase chain reaction (RT-qPCR) were used to detect the expression level of IL-1β, IL-6, TNF-α, miR-194 and CXCR4, respectively. In addition, the targeting relationship between miR-194 and CXCR4 was verified by dual-luciferase reporter gene assay. The dry/wet ratio of lung and the MPO activity were also measured to assess the inflammatory response in mice. Activation of NF-κB down-regulated the miR-194 expression in LPS-induced ALI. Overexpression of miR-194 alleviated LPS-induced ALI and reduced the expression of inflammatory factors IL-1β, IL-6 and TNF-α via targeting CXCR4. In LPS-induced ALI, NF-κB mediates the CXCR4 expression by inhibiting the expression of miR-194, thus promoting the inflammatory injury of lung.