MilliporeSigma
  • Evaluation of salivary and serum ADMA levels in patients with periodontal and cardiovascular disease as subclinical marker of cardiovascular risk.
Evaluation of salivary and serum ADMA levels in patients with periodontal and cardiovascular disease as subclinical marker of cardiovascular risk.
Journal of periodontology (2020-01-09)
Gaetano Isola, Angela Alibrandi, Monica Currò, Marco Matarese, Sergio Ricca, Giovanni Matarese, Riccardo Ientile, Thomas Kocher
ABSTRACT
Asymmetric dimethylarginine (ADMA) plays a crucial role in endothelial function and maybe a link for the known interaction of periodontitis and coronary heart disease (CHD). In this pilot study, we compared the impact of gingival health, periodontitis (CP), CHD, or of both diseases (CP + CHD) on salivary and serum ADMA levels. The clinical and periodontal characteristics, serum, and saliva samples were collected from 35 patients with CP, 33 patients with CHD, 35 patients with both CP + CHD, and 35 healthy subjects. Levels of ADMA and C-reactive protein (CRP) were assessed with a commercially available kit. The median (25% and 75% percentile) concentrations of salivary and serum ADMA were significantly higher in the CHD group [serum: 1.5 (1.2 to 1.8) μmol/L; salivary 1.3 (1 to 1.7) μmol/g protein, P < 0.01] and in the CP + CHD [serum: 1.8 (1.4 to 2.0) μmol/L; salivary 1.5 (1.2 to 1.7) μmol/g protein, P < 0.001] group compared to CP patients and controls. In univariate models, CP (P = 0.034), CHD (P < 0.001), and hs-CRP (P < 0.001) were significantly associated with serum ADMA, whereas in a multivariate model, hs-CRP remained a significant predictor of serum ADMA (P < 0.001). In a multivariate model, the significant predictors of salivary ADMA levels were hs-CRP (P < 0.001) and education socioeconomic status (P = 0.042). Patients with CHD and CP + CHD presented higher levels of salivary and serum ADMA compared to healthy subjects and CP patients. hs-CRP was a significant predictor of increased salivary and serum ADMA levels.
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride