MilliporeSigma
  • The Number of Transcription Factors at an Enhancer Determines Switch-like Gene Expression.
The Number of Transcription Factors at an Enhancer Determines Switch-like Gene Expression.
Cell reports (2020-06-04)
Hiroki Michida, Hiroaki Imoto, Hisaaki Shinohara, Noriko Yumoto, Masahide Seki, Mana Umeda, Tetsutaro Hayashi, Itoshi Nikaido, Takeya Kasukawa, Yutaka Suzuki, Mariko Okada-Hatakeyama
ABSTRACT
NF-κB is a transcription factor that activates super enhancers (SEs) and typical enhancers (TEs) and triggers threshold and graded gene expression, respectively. However, the mechanisms by which NF-κB selectively participates in these enhancers remain unclear. Here we show using mouse primary B lymphocytes that SE activity simultaneously associates with chromatin opening and enriched NF-κB binding, resulting in a higher fold change and threshold expression upon B cell receptor (BCR) activation. The higher fold change results from longer DNA, whereas the threshold response is explained by synergy in DNA-NF-κB binding and is supported by the coexistence of PU.1 and NF-κB in a SE before cell stimulation. This model indicates that the pre-existing NF-κB functions as a seed and triggers its processive binding upon BCR activation. Our mathematical modeling of the single-cell transcriptome reveals an additional role for SEs in divergent clonal responses in B cells.
Sigma-Aldrich
(+/-)-JQ1, ≥98% (HPLC)