MilliporeSigma
  • MiR-16-5p regulates postmenopausal osteoporosis by directly targeting VEGFA.
MiR-16-5p regulates postmenopausal osteoporosis by directly targeting VEGFA.
Aging (2020-05-20)
Tao Yu, Xiaomeng You, Haichao Zhou, Wenbao He, Zihua Li, Bing Li, Jiang Xia, Hui Zhu, Youguang Zhao, Guangrong Yu, Yuan Xiong, Yunfeng Yang
ABSTRACT
In this study, we used bioinformatics tools, and experiments with patient tissues and human mesenchymal stem cells (hMSCs) to identify differentially regulated genes (DEGs) and microRNAs (miRNAs) that promote postmenopausal osteoporosis. By analyzing the GSE56815 dataset from the NCBI GEO database, we identified 638 DEGs, including 371 upregulated and 267 downregulated genes, in postmenopausal women with low bone density. Enrichment and protein-protein interaction network analyses showed that TP53, RPS27A, and VEGFA were the top three hub genes with the highest degree of betweenness and closeness centrality. TargetScanHuman and DIANA software analyses and dual luciferase reporter assays confirmed that miR-16a-5p directly targets the 3'UTR of VEGFA. Postmenopausal patients with osteoporosis showed higher miR-16-5p and lower VEGFA levels than those without osteoporosis (n=10 each). VEGFA levels were higher in miR-16-5p knockdown hMSCs and were reduced in miR-16-5p-overexpressing hMSCs. mRNA expression of osteogenic markers, ALP, OCN, and RUNX2, as well as calcium deposition based on Alizarin red staining, correlated inversely with miR-16-5p levels and correlated positively with VEGFA levels. These findings suggest that miR-16-5p suppresses osteogenesis by inhibiting VEGFA expression and is a promising target for postmenopausal osteoporosis therapy.
Sigma-Aldrich
Anti-VEGFA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Pricing and availability is not currently available.
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Pricing and availability is not currently available.