MilliporeSigma
  • Separation of Zn(II), Cr(III), and Ni(II) Ions Using the Polymer Inclusion Membranes Containing Acetylacetone Derivative as the Carrier.
Separation of Zn(II), Cr(III), and Ni(II) Ions Using the Polymer Inclusion Membranes Containing Acetylacetone Derivative as the Carrier.
Membranes (2020-05-06)
Elzbieta Radzyminska-Lenarcik, Ilona Pyszka, Malgorzata Ulewicz
ABSTRACT
Polymer inclusion membranes (PIMs) doped with ethylenodiamino-bis-acetylacetone as fixed carrier was applied for the investigation of the facilitated transport of Zn(II), Cr(III), and Ni(II) ions from an aqueous nitrate feed phase (cM = 0.001 mol/dm3). The optimal membrane composition (amount of carrier and o-NPPE-plasticizer) was determined. For the optimal polymer inclusion membranes doped with ethylenodiamino-bis-acetylacetone, the following patterns of transport selectivity were found: Zn(II) > Cr(III) > Ni(II). The initial flux of Zn(II), Cr(III), and Ni(II) ions was 6.37 µmol/m2∙s, 5.53 µmol/m2∙s, and 0.40 µmol/m2∙s, respectively. The selectivity coefficients equal to 1.2 and 15.9 were found for Zn(II)/Cr(III) and Zn(II)/Ni(II), respectively. After 24-h transport, the recovery factor of Zn(II), Cr(III), and Ni(II) were 90%, 65%, and 6%, respectively. The polymer inclusion membranes doped with ethylenodiamino-bis-acetylacetone were characterized by scanning electron microscopy and non-contact atomic force microscopy. The influence of membrane morphology on transport process was discussed.