MilliporeSigma
  • Evaluation of pyraoxystrobin bioconcentration in zebrafish (Danio rerio) using modified QuEChERS extraction.
Evaluation of pyraoxystrobin bioconcentration in zebrafish (Danio rerio) using modified QuEChERS extraction.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2020-05-26)
Xiaohong Yang, Huijun Liu, Minghong Jia, Jiangfei Wang, Jinlong Wu, Junhua Song, Yixuan Liu
ABSTRACT
Pyraoxystrobin is a novel strobilurin fungicide that is widely used on many crops. The high log Kow of pyraoxystrobin implies that it tends to accumulate in aquatic organisms. This study optimized the sorbents of QuEChERS (quick, easy, cheap, effective, rugged, and safe) using 13C-labelled pyraoxystrobin as the internal standard (IS). It has been established a QuEChERS-LC-MS/MS IS method to study the bioconcentration and elimination of pyraoxystrobin in zebrafish (Danio rerio). The results indicated that the method had satisfactory linearity between 0.234 and 15 μg L-1 (R2 = 0.9996). The limits of detection (LOD) and quantification (LOQ) for pyraoxystrobin were 0.01 and 0.03 μg L-1, respectively. The LOQs of the method for water and zebrafish were 0.05 μg L-1 and 0.01 mg/kg, respectively. The mean recovery of pyraoxystrobin in zebrafish and water at fortification levels of 0.01-0.3 mg kg-1 and 0.05-1.5 μg L-1 ranged from 98.31 to 105.61% and 101.87 to 108.48%, respectively, with a % RSD (relative standard deviation) of 0.94-3.57%. The bioconcentration has been evaluated. The bioconcentration factors for pyraoxystrobin in zebrafish were 1,792 and 3,505 after exposure to 0.5 μg L-1 for 168 h and 0.05 μg L-1 for 216 h, respectively. The half-life of pyraoxystrobin in zebrafish was 9.0-9.5 d.
Sigma-Aldrich
N-Propylethylenediamine, ≥97%