MilliporeSigma
  • Inhibitory Effects of AF-343, a Mixture of Cassia tora L., Ulmus pumila L., and Taraxacum officinale, on Compound 48/80-Mediated Allergic Responses in RBL-2H3 Cells.
Inhibitory Effects of AF-343, a Mixture of Cassia tora L., Ulmus pumila L., and Taraxacum officinale, on Compound 48/80-Mediated Allergic Responses in RBL-2H3 Cells.
Molecules (Basel, Switzerland) (2020-05-28)
Eun Kyeong Lee, Jeongah Song, Youjin Seo, Eun Mi Koh, Seon-Hee Kim, Kyung Jin Jung
ABSTRACT
The purpose of this study was to determine the antiallergic effects of AF-343, a mixture of natural plant extracts from Cassia tora L., Ulmus pumila L., and Taraxacum officinale, on rat basophilic leukemia (RBL-2H3) cells. The inhibitory effects on cell degranulation, proinflammatory cytokine secretion, and reactive oxygen species (ROS) production were studied in compound 48/80-treated RBL-2H3 cells. The bioactive compounds in AF-343 were also identified by HPLC-UV. AF-343 was found to effectively suppress compound 48/80-induced b-hexosaminidase release, and interleukin (IL)-4 and tumor necrosis factor-a (TNF-a) production in RBL-2H3 cells. In addition, AF-343 exhibited DPPH free radical scavenging effects in vitro (half-maximal inhibitory concentration (IC50) = 105 μg/mL) and potently inhibited compound 48/80-induced cellular ROS generation in a 2',7'-dichlorofluorescein diacetate (DCFH-DA) assay. Specifically, treatment with AF-343 exerted stronger antioxidant effects in vitro and antiallergic effects in cells than treatment with three single natural plant extracts. Furthermore, AF-343 was observed to contain bioactive compounds, including catechin, aurantio-obtusin, and chicoric acid, which have been reported to elicit antiallergic responses. This study reveals that AF-343 attenuates allergic responses via suppression of b-hexosaminidase release, IL-4 and TNF-a secretion, and ROS generation, perhaps through mechanisms related to catechin, aurantio-obtusin, and chicoric acid. The results indicate that AF-343 can be considered a treatment for various allergic diseases.
Product Description
Aurantio-obtusin, phyproof® Reference Substance