MilliporeSigma
  • Is anti-h-caldesmon useful for distinguishing smooth muscle and myofibroblastic tumors? An immunohistochemical study.
Is anti-h-caldesmon useful for distinguishing smooth muscle and myofibroblastic tumors? An immunohistochemical study.
American journal of clinical pathology (2000-11-09)
K M Ceballos, G P Nielsen, M K Selig, J X O'Connell
ABSTRACT
Misinterpretation of positive staining of antibodies to desmin, smooth muscle actin, and muscle actin as representing smooth muscle differentiation in the context of a spindle cell tumor is not uncommon. Anti-h-caldesmon is a promising novel immunohistochemical reagent for more specific smooth muscle differentiation. We studied 72 tumors (11 leiomyosarcomas, 26 malignant fibrous histiocytomas [MFHs], 11 fibromatoses, 11 cellular cutaneous fibrous histiocytomas [CCFHs], 5 malignant peripheral nerve sheath tumors, 4 synovial sarcomas, and 4 cases of nodular fasciitis), the reactive myofibroblastic response in 5 cases of acute cholecystitis, and the desmoplastic response surrounding 5 invasive breast carcinomas. Tissues were examined for expression of h-caldesmon, desmin, smooth muscle actin, and muscle actin. Diffuse staining for h-caldesmon was present only within the leiomyosarcomas. Focal staining for h-caldesmon involving less than 1% of lesional cells was present in 3 of 26 MFHs and 1 of 11 CCFHs. There was overlap in staining for the other "myoid" markers in all of the lesions that contained myofibroblasts. Anti-h-caldesmon seems to be a reliable marker of smooth muscle differentiation, and its inclusion in a panel of myoid immunohistochemical reagents should allow distinction of smooth muscle and myofibroblastic tumors.