MilliporeSigma
  • Non-clinical pharmacokinetic profiles of rovatirelin, an orally available thyrotropin-releasing hormone analogue.
Non-clinical pharmacokinetic profiles of rovatirelin, an orally available thyrotropin-releasing hormone analogue.
Xenobiotica; the fate of foreign compounds in biological systems (2018-01-05)
Kaoru Kobayashi, Yoshikazu Abe, Hiroshi Harada, Emiko Oota, Takuro Endo, Hiroo Takeda
ABSTRACT
1. The non-clinical pharmacokinetic profiles of rovatirelin, a novel thyrotropin-releasing hormone (TRH) analogue, were investigated in vivo and in vitro. 2. Rovatirelin orally administered to rats and dogs was rapidly absorbed and bioavailability was estimated to be 7.3 and 41.3%, respectively. The extent of plasma protein binding of rovatirelin in rats, dogs, and humans was low in all species (∼15%). The permeability of rovatirelin from blood to brain (permeability-surface area) ranged from 1.04 ± 0.14 to 1.29 ± 0.28 μL/min/g in rats, and rovatirelin was stable in rat plasma and brain homogenates. 3. The metabolite pattern was qualitatively similar in vitro and in vivo. In animals, rovatirelin aminopentanoic acid (rovatirelin-acid), rovatirelin aminopentanone (rovatirelin-ketone), rovatirelin pyrrolidine (4S)-hydroxy (rovatirelin-OH), (thiazoylalanyl)methylpyrrolidine (TAMP), 3-(4-thiazoyl)-l-alanine (TA), and unknown metabolites were observed. In human hepatocytes, TAMP was mainly formed and no unique human metabolite was observed. 4. The radioactivity from administered [14C]rovatirelin was predominantly excreted in faeces in rats and dogs, and almost all radioactivity was recovered 168 h after administration. Absorption, brain penetration, and stability of rovatirelin in the brain were greater than for taltirelin. 5. Thus, orally administered rovatirelin is a potentially improved treatment for spinocerebellar degeneration compared with taltirelin.
Sigma-Aldrich
Rovatirelin, ≥98% (HPLC)