MilliporeSigma
SUMO1 haploinsufficiency leads to cleft lip and palate.
Science (New York, N.Y.) (2006-09-23)
Fowzan S Alkuraya, Irfan Saadi, Jennifer J Lund, Annick Turbe-Doan, Cynthia C Morton, Richard L Maas
ABSTRACT
The posttranslational modification sumoylation can have multiple effects on its substrate proteins. We studied a patient with isolated cleft lip and palate and a balanced chromosomal translocation that disrupts the SUMO1 (small ubiquitin-related modifier) gene, resulting in haploinsufficiency. In mouse, we found that Sumo1 is expressed in the developing lip and palate and that a Sumo1 hypomorphic allele manifests an incompletely penetrant orofacial clefting phenotype. Products of several genes implicated in clefting are sumoylated, and the Sumo1 hypomorphic allele interacts genetically with a loss-of-function allele for one of these loci. Thus, sumoylation defines a network of genes important for palatogenesis.