MilliporeSigma
  • Effects of donor age on neoplastic transformation of adult mouse bladder epithelium in vitro.
Effects of donor age on neoplastic transformation of adult mouse bladder epithelium in vitro.
Journal of the National Cancer Institute (1979-04-01)
I C Summerhayes, L M Franks
PMID107359
ABSTRACT
Neoplastic transformation of C57BL/lcrf-a' mouse bladder epithelium was induced in long-term primary cultures by a single 24-hour treatment with 7,12-dimethylbenz[a]anthracene on day 2 of culture. Transformed foci appeared earlier (40--60 days) and at a higher frequency (28%) in cultures from old donors (28--30 mo) compared with 100 days and 0.9% in cultures from young adult donors (5--7 mo). After transplantation into syngeneic mice, transformed cells produced carcinomas. Spontaneous epithelial transformation occurred in dimethyl sulfoxide-treated old cultures after the same interval (40--60 days) as in the carcinogen-treated cultures but at a lower frequency (5.3%). Spontaneous epithelial transformation did not occur in cultures from young donors.
Sigma-Aldrich
MB49 Mouse Bladder Carcinoma Cell Line, MB49 mouse urothelial carcinoma cell line is widely used as an in vitro and in vivo model of bladder cancer.