MilliporeSigma
  • The renoprotective effects of mannitol and udenafil in renal ischemia-reperfusion injury model.
The renoprotective effects of mannitol and udenafil in renal ischemia-reperfusion injury model.
Investigative and clinical urology (2017-07-07)
Yusuf Özlülerden, Cihan Toktaş, Hülya Aybek, Vural Küçükatay, Nilay Şen Türk, Ali Ersin Zumrutbas
ABSTRACT
The aim of this study was to investigate and compare the effects of udenafil and mannitol in an experimental renal ischemia-reperfusion (I/R) injury model. A total of 64 female Wister Albino rats were used. Right nephrectomy was performed in all groups. In the control group; I/R injury was not performed. In the I/R group; left renal pedicle was clamped for 45 minutes and then underwent 60 minutes and 24 hours of reperfusion. In the mannitol group; 1 mL 20% mannitol was given intravenously 15 minutes before clamping. In the udenafil group; 10-mg/kg udenafil was given orally 1 hour before clamping. Creatinine (Cr), blood urea nitrogen (BUN), Cr clearance, malondialdehyde, neutrophil gelatinase associated lipocalin (NGAL), histological examination and DNA damage (Comet Assay method) levels were compared in tissue, serum and urine samples. Udenafil had a better protective effect than mannitol according to biochemical parameters (Cr, BUN, Cr clearance, and NGAL levels) and histopathological findings when compared with the I/R group. In the Comet sampling analysis no significant difference was detected. Udenafil has a better renoprotective effect than mannitol against I/R injury and this effect supports more functional improvements. Further clinical trials are needed to demonstrate those effects and clinical utility of udenafil for that purpose in humans.
Sigma-Aldrich
Udenafil, ≥98% (HPLC)