MilliporeSigma
  • A general analytical strategy for the characterization of phenolic compounds in fruit juices by high-performance liquid chromatography with diode array detection coupled to electrospray ionization and triple quadrupole mass spectrometry.
A general analytical strategy for the characterization of phenolic compounds in fruit juices by high-performance liquid chromatography with diode array detection coupled to electrospray ionization and triple quadrupole mass spectrometry.
Journal of chromatography. A (2009-06-09)
Beatriz Abad-García, Luis A Berrueta, Sergio Garmón-Lobato, Blanca Gallo, Francisca Vicente
ABSTRACT
In the present study, a methodology based on liquid chromatography with diode array detection (HPLC/DAD) coupled to an electrospray ionization (ESI) interface and a triple quadrupole mass spectrometer for the simultaneous identification of phenolic compounds in fruit juices has been developed. 72 available phenolic compound standards from diverse families present in fruits have been studied in order to analyze their fragmentation pattern. As a result, a general strategy for the characterization of unknown phenolic compounds in fruit juices was designed: (i) taking into account its UV-visible spectrum and elution order, assign the unknown polyphenol to a polyphenol class, (ii) identify the quasi-molecular ion using positive and negative MS spectra, being supported by adducts generated with solvent or sodium and molecular complexes, (iii) determinate the pattern of glycosylation in positive mode using ESI(+)-CID MS/MS product ion scan experiments, selecting the quasi-molecular ion as precursor ion, and finally, (iv) study the identity of the aglycone through ESI(+)-CID MS/MS product ion spectra from the protonated aglycone, [Y(0)](+). This strategy was successfully employed for the characterization of known and unknown phenolic compounds in juices from 17 different fruits.