MilliporeSigma
  • A Designed Peptide Targets Two Types of Modifications of p53 with Anti-cancer Activity.
A Designed Peptide Targets Two Types of Modifications of p53 with Anti-cancer Activity.
Cell chemical biology (2018-04-24)
Lunxi Liang, Huanbin Wang, Hubing Shi, Zhaoli Li, Han Yao, Zhigao Bu, Ningning Song, Chushu Li, Dabin Xiang, Yao Zhang, Jilin Wang, Ye Hu, Qi Xu, Yanlei Ma, Zhongyi Cheng, Yingchao Wang, Shuliang Zhao, Jin Qian, Yingxuan Chen, Jing-Yuan Fang, Jie Xu
ABSTRACT
Many cancer-related proteins are controlled by composite post-translational modifications (PTMs), but prevalent strategies only target one type of modification. Here we describe a designed peptide that controls two types of modifications of the p53 tumor suppressor, based on the discovery of a protein complex that suppresses p53 (suppresome). We found that Morn3, a cancer-testis antigen, recruits different PTM enzymes, such as sirtuin deacetylase and ubiquitin ligase, to confer composite modifications on p53. The molecular functions of Morn3 were validated through in vivo assays and chemico-biological intervention. A rationally designed Morn3-targeting peptide (Morncide) successfully activated p53 and suppressed tumor growth. These findings shed light on the regulation of protein PTMs and present a strategy for targeting two modifications with one molecule.
Sigma-Aldrich
SAR405838, ≥98% (HPLC)