MilliporeSigma
  • Jdp2-deficient granule cell progenitors in the cerebellum are resistant to ROS-mediated apoptosis through xCT/Slc7a11 activation.
Jdp2-deficient granule cell progenitors in the cerebellum are resistant to ROS-mediated apoptosis through xCT/Slc7a11 activation.
Scientific reports (2020-03-20)
Chia-Chen Ku, Kenly Wuputra, Kohsuke Kato, Wen-Hsin Lin, Jia-Bin Pan, Shih-Chieh Tsai, Che-Jung Kuo, Kan-Hung Lee, Yan-Liang Lee, Ying-Chu Lin, Shigeo Saito, Michiya Noguchi, Yukio Nakamura, Hiroyuki Miyoshi, Richard Eckner, Kyosuke Nagata, Deng-Chyang Wu, Chang-Shen Lin, Kazunari K Yokoyama
ABSTRACT
The Jun dimerization protein 2 (Jdp2) is expressed predominantly in granule cell progenitors (GCPs) in the cerebellum, as was shown in Jdp2-promoter-Cre transgenic mice. Cerebellum of Jdp2-knockout (KO) mice contains lower number of Atoh-1 positive GCPs than WT. Primary cultures of GCPs from Jdp2-KO mice at postnatal day 5 were more resistant to apoptosis than GCPs from wild-type mice. In Jdp2-KO GCPs, the levels of both the glutamate‒cystine exchanger Sc7a11 and glutathione were increased; by contrast, the activity of reactive oxygen species (ROS) was decreased; these changes confer resistance to ROS-mediated apoptosis. In the absence of Jdp2, a complex of the cyclin-dependent kinase inhibitor 1 (p21Cip1) and Nrf2 bound to antioxidant response elements of the Slc7a11 promoter and provide redox control to block ROS-mediated apoptosis. These findings suggest that an interplay between Jdp2, Nrf2, and p21Cip1 regulates the GCP apoptosis, which is one of critical events for normal development of the cerebellum.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Anti-ATH 1 Antibody, Chemicon®, from rabbit