MilliporeSigma
  • Paradoxical activation of the protein kinase-transcription factor ERK5 by ERK5 kinase inhibitors.
Paradoxical activation of the protein kinase-transcription factor ERK5 by ERK5 kinase inhibitors.
Nature communications (2020-03-15)
Pamela A Lochhead, Julie A Tucker, Natalie J Tatum, Jinhua Wang, David Oxley, Andrew M Kidger, Victoria P Johnson, Megan A Cassidy, Nathanael S Gray, Martin E M Noble, Simon J Cook
ABSTRACT
The dual protein kinase-transcription factor, ERK5, is an emerging drug target in cancer and inflammation, and small-molecule ERK5 kinase inhibitors have been developed. However, selective ERK5 kinase inhibitors fail to recapitulate ERK5 genetic ablation phenotypes, suggesting kinase-independent functions for ERK5. Here we show that ERK5 kinase inhibitors cause paradoxical activation of ERK5 transcriptional activity mediated through its unique C-terminal transcriptional activation domain (TAD). Using the ERK5 kinase inhibitor, Compound 26 (ERK5-IN-1), as a paradigm, we have developed kinase-active, drug-resistant mutants of ERK5. With these mutants, we show that induction of ERK5 transcriptional activity requires direct binding of the inhibitor to the kinase domain. This in turn promotes conformational changes in the kinase domain that result in nuclear translocation of ERK5 and stimulation of gene transcription. This shows that both the ERK5 kinase and TAD must be considered when assessing the role of ERK5 and the effectiveness of anti-ERK5 therapeutics.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-MEK5 Antibody, Chemicon®, from rabbit