MilliporeSigma
  • [18F]azadibenzocyclooctyne ([18F]ADIBO): a biocompatible radioactive labeling synthon for peptides using catalyst free [3+2] cycloaddition.
[18F]azadibenzocyclooctyne ([18F]ADIBO): a biocompatible radioactive labeling synthon for peptides using catalyst free [3+2] cycloaddition.
Bioorganic & medicinal chemistry letters (2011-10-26)
Selvanathan Arumugam, Joshua Chin, Ralf Schirrmacher, Vladimir V Popik, Alexey P Kostikov
ABSTRACT
N-Terminally azido-modified peptides were labeled with the novel prosthetic labeling synthon [(18)F]azadibenzocyclooctyne ([(18)F]ADIBO) using copper-free azide-alkyne [3+2]-dipolar cycloaddition in high radiochemical yields (RCYs). (18)F-Labeled [(18)F]ADIBO was prepared by nucleophilic substitution of the corresponding tosylate in 21% overall RCY (EOB) in a fully automated synthesis unit within 55 min. [(18)F]ADIBO was incubated with azide-containing peptides at room temperature in the absence of toxic metal catalysts and the formation of the triazole conjugate was confirmed. Finally, the azide-alkyne [3+2]-dipolar cycloaddition was shown to proceed with 95% radiochemical yield in ethanol within 30 min, allowing for a development of a kit-like peptide labeling approach with [(18)F]ADIBO.
Sigma-Aldrich
Dibenzocyclooctyne-acid, 95%, storage temp.:-20°C
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-acid, for Copper-free Click Chemistry